Calcium, 99%, turnings, Thermo Scientific Chemicals
Calcium, 99%, turnings, Thermo Scientific Chemicals
Calcium, 99%, turnings, Thermo Scientific Chemicals
Calcium, 99%, turnings, Thermo Scientific Chemicals
Calcium, 99%, turnings, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA365741000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium
ColorColorless to Yellow
Name Note99%
CAS7440-70-2
Health Hazard 1GHS Signal Word: Danger