Lithium metaborate, 99%, for analysis
Lithium metaborate, 99%, for analysis
Lithium metaborate, 99%, for analysis
Lithium metaborate, 99%, for analysis
Thermo Scientific Chemicals
Lithium metaborate, 99%, for analysis
CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA365715000
Price (MYR)
3,558.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,558.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLithium metaborate
Melting Point845°C
Name Note99%
CAS13453-69-5