2-Bromo-4'-cyanoacetophenone, 97%
2-Bromo-4'-cyanoacetophenone, 97%
2-Bromo-4'-cyanoacetophenone, 97%
2-Bromo-4'-cyanoacetophenone, 97%
Thermo Scientific Chemicals
2-Bromo-4'-cyanoacetophenone, 97%
CAS: 20099-89-2 | C9H6BrNO | 224.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA365360250
Price (MYR)
1,014.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,014.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromo-4'-cyanoacetophenone
Name Note97%
CAS20099-89-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger