Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Thermo Scientific Chemicals
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
CAS: 34513-98-9 | N4O10Ru | 317.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA365045000
Price (MYR)
14,526.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
14,526.00
EA
Add to cart
Specifications
Chemical Name or MaterialRuthenium(III) nitrosyl nitrate
Name NoteIn diluted nitric acid
CAS34513-98-9
ColorBrown to Orange
Health Hazard 1GHS Signal Word: Danger