Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Thermo Scientific Chemicals
Potassium tert-butoxide, pure, 20 wt.% solution in THF
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA364990500
Price (MYR)
404.00
BOT
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
404.00
BOT
Add to cart
Chemical Identifiers
CAS865-47-4, 109-99-9
Specifications
Chemical Name or MaterialPotassium tert-butoxide
CAS865-47-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapour.
Suspected of causing cancer.
Reacts violently with water.
May form explosive peroxides.<br
CAS109-99-9