Benzyltrimethylammonium bromide, 98+%
Benzyltrimethylammonium bromide, 98+%
Benzyltrimethylammonium bromide, 98+%
Benzyltrimethylammonium bromide, 98+%
Thermo Scientific Chemicals
Benzyltrimethylammonium bromide, 98+%
CAS: 5350-41-4 | C10H16BrN | 230.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA364860250
Price (MYR)
246.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
246.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyltrimethylammonium bromide
Name Note99%
CAS5350-41-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
View more
Phase transfer catalyst
RUO – Research Use Only