Guanidine hydrochloride, 99.5%, without anticaking agent
Guanidine hydrochloride, 99.5%, without anticaking agent
Guanidine hydrochloride, 99.5%, without anticaking agent
Guanidine hydrochloride, 99.5%, without anticaking agent
Thermo Scientific Chemicals
Guanidine hydrochloride, 99.5%, without anticaking agent
CAS: 50-01-1 | CH5N3·HCl | 95.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA364795000
Price (MYR)
1,136.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,136.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGuanidine hydrochloride
Name NoteWithout anticaking agent
CAS50-01-1
Absorbance(6M aq. soln.), (6 M aq. soln.) at 0nm, 0.01 max. at 280nm, 0.03 max. at 260nm
Health Hazard 1GHS Signal Word: Warning