Methyl sulfoxide-d6, for NMR, 99.8 atom % D
Methyl sulfoxide-d6, for NMR, 99.8 atom % D
Methyl sulfoxide-d6, for NMR, 99.8 atom % D
Methyl sulfoxide-d6, for NMR, 99.8 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99.8 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
10 mL
25 mL
50 mL
Packaging:
Glass Bottle
Catalog number FSA364651000
Price (MYR)
2,363.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,363.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Density1.1900g/mL
Deuterium (D)99.75% min.
Flash Point88°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only