Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA364350025
Price (MYR)
1,213.00
LT
Add to cart
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,213.00
LT
Add to cart
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
Name NoteOver Molecular Sieves
CAS141-78-6