Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Thermo Scientific Chemicals
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA364230025
Price (MYR)
677.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
677.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
Name NoteStabilized
Absorbance(1 cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.02 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm
CAS75-09-2
View more
for HPLC
RUO – Research Use Only