(S)-N-BOC-Propargylglycine, 95%
(S)-N-BOC-Propargylglycine, 95%
(S)-N-BOC-Propargylglycine, 95%
(S)-N-BOC-Propargylglycine, 95%
Thermo Scientific Chemicals
(S)-N-BOC-Propargylglycine, 95%
CAS: 63039-48-5 | C10H15NO4 | 213.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA361800010
Price (MYR)
1,262.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,262.00
BOT
Add to cart
Specifications
Chemical Name or Material(S)-N-BOC-Propargylglycine
Name Note95%, (98% e.e.)
CAS63039-48-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning