Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Thermo Scientific Chemicals
Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
CAS: 71889-02-6 | C18H37Cl3O2Si2 | 448.012 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass Bottle
Catalog number FSA360122500
Price (MYR)
4,102.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
4,102.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilica gel, C18-RP, 17%C
CAS71889-02-6
Quantity250 g
PackagingGlass Bottle
Molecular FormulaC18H37Cl3O2Si2
View more
For column chromatography
RUO – Research Use Only