Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron
Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron
Thermo Scientific Chemicals
Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number FSA360112500
Price (MYR)
6,188.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
6,188.00
EA
Add to cart
Specifications
PackagingGlass Bottle
Quantity250 g
MaterialSilica Gel
Unit SizeEA
For column chromatography
RUO – Research Use Only