Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Thermo Scientific Chemicals
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA360062500
Price (MYR)
383.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
383.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSilica gel
ColorWhite to Brown
Physical FormPowder and/or Chunks
pH6.5 to 7.5 (10% w/w suspension)
Name NoteFor column chromatography
View more
For column chromatography
RUO – Research Use Only