Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Thermo Scientific Chemicals
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
For column chromatography. | CAS: 7631-86-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number FSA360050050
Price (MYR)
3,513.00
BOT
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
3,513.00
BOT
Add to cart
Specifications
Particle Size40 to 60 μm
Pore Size60 Å
PackagingPlastic Drum
Quantity5 kg
TypeSilica Gel