Silica gel, for column chromatography,ultra pure, 40-63μm, 40A
Silica gel, for column chromatography,ultra pure, 40-63μm, 40A
Silica gel, for column chromatography,ultra pure, 40-63μm, 40A
Silica gel, for column chromatography,ultra pure, 40-63μm, 40A
Silica gel, for column chromatography,ultra pure, 40-63μm, 40A
Thermo Scientific Chemicals
Silica gel, for column chromatography,ultra pure, 40-63μm, 40A
CAS: 7631-86-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA360031000
Price (MYR)
409.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
409.00
BOT
Add to cart
Specifications
Particle Size40 to 60 μm
Pore Size40 Å
PackagingPlastic Bottle
Quantity100 g
TypeSilica Gel
View more
For column chromatography
RUO – Research Use Only