Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals
Lithium trifluoromethanesulfonate, 98%
CAS: 33454-82-9 | CF3LiO3S | 156.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA358711000
Price (MYR)
2,443.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,443.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLithium trifluoromethanesulfonate
Name Note98%
CAS33454-82-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Suitable for battery materials development.
RUO – Research Use Only