Dipentyl phthalate, 97%
Dipentyl phthalate, 97%
Dipentyl phthalate, 97%
Dipentyl phthalate, 97%
Thermo Scientific Chemicals
Dipentyl phthalate, 97%
CAS: 131-18-0 | C18H26O4 | 306.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA357361000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDipentyl phthalate
CAS131-18-0
Health Hazard 1GHS Signal Word: Danger
Melting Point-55.0°C
ColorColorless to Yellow