1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%
Thermo Scientific Chemicals
1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%
CAS: 304680-36-2 | C12H23F6N2P | 340.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA356760050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Octyl-3-methylimidazolium hexafluorophosphate
CAS304680-36-2
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Ionic liquid
RUO – Research Use Only