1-Ethyl-3-methylimidazolium bromide, 98%
1-Ethyl-3-methylimidazolium bromide, 98%
1-Ethyl-3-methylimidazolium bromide, 98%
1-Ethyl-3-methylimidazolium bromide, 98%
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium bromide, 98%
CAS: 65039-08-9 | C6H11BrN2 | 191.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA356671000
Price (MYR)
1,642.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,642.00
EA
Add to cart
Specifications
Chemical Name or Material1-Ethyl-3-methylimidazolium bromide
CAS65039-08-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Ionic liquid
RUO – Research Use Only