n-Nonanoic acid, 97%
n-Nonanoic acid, 97%
n-Nonanoic acid, 97%
n-Nonanoic acid, 97%
Thermo Scientific Chemicals
n-Nonanoic acid, 97%
CAS: 112-05-0 | C9H18O2 | 158.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA356241000
Price (MYR)
192.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
192.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Nonanoic acid
CAS112-05-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0