Phorbol 12-myristate 13-acetate, 97%
Phorbol 12-myristate 13-acetate, 97%
Phorbol 12-myristate 13-acetate, 97%
Phorbol 12-myristate 13-acetate, 97%
Thermo Scientific Chemicals
Phorbol 12-myristate 13-acetate, 97%
CAS: 16561-29-8 | C36H56O8 | 616.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
1 mg
Packaging:
Glass bottle
Catalog number FSA356150050
Price (MYR)
2,372.00
EA
Add to cart
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,372.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhorbol 12-myristate 13-acetate
CAS16561-29-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a