2,7-Dibromofluorene, 99%
2,7-Dibromofluorene, 99%
2,7-Dibromofluorene, 99%
2,7-Dibromofluorene, 99%
Thermo Scientific Chemicals
2,7-Dibromofluorene, 99%
CAS: 16433-88-8 | C13H8Br2 | 324.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA355240250
Price (MYR)
1,209.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,209.00
EA
Add to cart
Specifications
Chemical Name or Material2, 7-Dibromofluorene
CAS16433-88-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0