n-Octadecyl mercaptan, 96%
n-Octadecyl mercaptan, 96%
n-Octadecyl mercaptan, 96%
n-Octadecyl mercaptan, 96%
Thermo Scientific Chemicals
n-Octadecyl mercaptan, 96%
CAS: 2885-00-9 | C18H38S | 286.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA354981000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Octadecyl mercaptan
CAS2885-00-9
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0