1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA354900025
Price (MYR)
1,330.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,330.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
CAS872-50-4
Health Hazard 1GHS Signal Word: Danger
Density1.0300g/mL
View more
Suitable for battery materials development.
RUO – Research Use Only