N,N-Dimethylformamide, 99.8%, for peptide synthesis
N,N-Dimethylformamide, 99.8%, for peptide synthesis
N,N-Dimethylformamide, 99.8%, for peptide synthesis
N,N-Dimethylformamide, 99.8%, for peptide synthesis
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, for peptide synthesis
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
Packaging:
Plastic Drum
Catalog number FSA354830100
Price (MYR)
3,083.00
EA
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
3,083.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
CAS68-12-2
CAS Min %99.75
ColorWhite
View more
For peptide synthesis
RUO – Research Use Only