Sodium sulfate, 99+%, ACS reagent, granular, anhydrous
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous
Thermo Scientific Chemicals
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous
CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic bottle
Catalog number FSA354250025
Price (MYR)
961.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
961.00
EA
Add to cart
Specifications
Boiling Point>1700.0°C
Chemical Name or MaterialSodium sulfate
Melting Point884.0°C
Name NoteAnhydrous
CAS7757-82-6