1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA354211000
Price (MYR)
1,980.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,980.00
EA
Add to cart
Specifications
Chemical Name or Material1-Butyl-3-methylimidazolium tetrafluoroborate
CAS174501-65-6
CAS Min %98.0
Health Hazard 1HYGROSCOPIC
CAS Max %100.0
View more
Ionic liquid
RUO – Research Use Only