1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA354171000
Price (MYR)
2,372.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,372.00
EA
Add to cart
Specifications
Chemical Name or Material1-Butyl-3-methylimidazolium hexafluorophosphate
CAS174501-64-5
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Ionic liquid
RUO – Research Use Only