4-Methoxyphenethyl alcohol, 96%
4-Methoxyphenethyl alcohol, 96%
4-Methoxyphenethyl alcohol, 96%
4-Methoxyphenethyl alcohol, 96%
Thermo Scientific Chemicals
4-Methoxyphenethyl alcohol, 96%
CAS: 702-23-8 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA354050100
Price (MYR)
688.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
688.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methoxyphenethyl alcohol
Name Note96%
CAS702-23-8
CAS Min %95.0
CAS Max %100.0