Tetrabutylammonium tetrafluoroborate, 98%
Tetrabutylammonium tetrafluoroborate, 98%
Tetrabutylammonium tetrafluoroborate, 98%
Tetrabutylammonium tetrafluoroborate, 98%
Thermo Scientific Chemicals
Tetrabutylammonium tetrafluoroborate, 98%
CAS: 429-42-5 | C16H36BF4N | 329.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number FSA353091000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrabutylammonium tetrafluoroborate
Name Note95%
CAS429-42-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Phase transfer catalyst
RUO – Research Use Only