Isophorondiamine, 99+%, mixture of cis and trans
Isophorondiamine, 99+%, mixture of cis and trans
Isophorondiamine, 99+%, mixture of cis and trans
Isophorondiamine, 99+%, mixture of cis and trans
Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans
CAS: 2855-13-2 | C10H22N2 | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA352942500
Price (MYR)
376.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
376.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsophorondiamine
Name Note99+%
CAS2855-13-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful to aquatic life with long lasting effects.