n-Heneicosane, 98%
n-Heneicosane, 98%
n-Heneicosane, 98%
Thermo Scientific Chemicals
n-Heneicosane, 98%
CAS: 629-94-7 | C21H44 | 296.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA352770100
Price (MYR)
1,084.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,084.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Heneicosane
CAS629-94-7
CAS Min %97.5
CAS Max %100.0
Melting Point39°C to 43°C