Pyridoxal hydrochloride, 99%
Pyridoxal hydrochloride, 99%
Pyridoxal hydrochloride, 99%
Pyridoxal hydrochloride, 99%
Thermo Scientific Chemicals
Pyridoxal hydrochloride, 99%
CAS: 65-22-5 | C8H9NO3·HCl | 203.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Packaging:
Glass bottle
Catalog number FSA352710250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPyridoxal hydrochloride
CAS65-22-5
Melting Point171.0°C to 175.0°C
ColorWhite
EINECS Number200-602-5