p-Anisaldehyde dimethyl acetal, 98%
p-Anisaldehyde dimethyl acetal, 98%
p-Anisaldehyde dimethyl acetal, 98%
p-Anisaldehyde dimethyl acetal, 98%
Thermo Scientific Chemicals
p-Anisaldehyde dimethyl acetal, 98%
CAS: 2186-92-7 | C10H14O3 | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA352291000
Price (MYR)
1,095.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,095.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Anisaldehyde dimethyl acetal
CAS2186-92-7
ColorColorless to Yellow
Density1.0680g/mL
EINECS Number218-577-4