Oxalic acid, potassium salt dihydrate, 99%, extra pure
Oxalic acid, potassium salt dihydrate, 99%, extra pure
Oxalic acid, potassium salt dihydrate, 99%, extra pure
Oxalic acid, potassium salt dihydrate, 99%, extra pure
Thermo Scientific Chemicals
Oxalic acid, potassium salt dihydrate, 99%, extra pure
CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA352115000
Price (MYR)
1,271.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,271.00
EA
Add to cart
Specifications
Chemical Name or MaterialOxalic acid, potassium salt dihydrate
Name Note99%
CAS6100-20-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement