N-(3-Aminopropyl)imidazole, 98%
N-(3-Aminopropyl)imidazole, 98%
N-(3-Aminopropyl)imidazole, 98%
N-(3-Aminopropyl)imidazole, 98%
Thermo Scientific Chemicals
N-(3-Aminopropyl)imidazole, 98%
CAS: 5036-48-6 | C6H11N3 | 125.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA351942500
Price (MYR)
1,251.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,251.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-(3-Aminopropyl)imidazole
Name Note98%
CAS5036-48-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger