Isosorbide, 98%
Isosorbide, 98%
Isosorbide, 98%
Thermo Scientific Chemicals
Isosorbide, 98%
CAS: 652-67-5 | C6H10O4 | 146.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA351911000
Price (MYR)
584.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
584.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsosorbide
Melting Point56.0°C to 62.0°C
CAS652-67-5
ColorBeige or White to Yellow
EINECS Number211-492-3