Sodium glycolate, 97%
Sodium glycolate, 97%
Sodium glycolate, 97%
Sodium glycolate, 97%
Thermo Scientific Chemicals
Sodium glycolate, 97%
CAS: 2836-32-0 | C2H3NaO3 | 98.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA351571000
Price (MYR)
447.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
447.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium glycolate
Name Note97%
CAS2836-32-0
ColorWhite
RTECS NumberMC5525000