4-Fluorophenethylamine, 99%
4-Fluorophenethylamine, 99%
4-Fluorophenethylamine, 99%
4-Fluorophenethylamine, 99%
Thermo Scientific Chemicals
4-Fluorophenethylamine, 99%
CAS: 1583-88-6 | C8H10FN | 139.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA350950050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Fluorophenethylamine
Name Note99%
CAS1583-88-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger