Check Order Status
2-Amino-5-bromobenzonitrile, 97%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
2-Amino-5-bromobenzonitrile, 97%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
Packaging:
Glass Bottle
Catalog number
FSA350250010
Price (MYR)
305.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
305.00
BOT
Add to cart
Specifications
Chemical Name or Material
2-Amino-5-bromobenzonitrile
CAS
39263-32-6
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0