Drierite™, with indicator, 10-20 mesh
Drierite™, with indicator, 10-20 mesh
Drierite™, with indicator, 10-20 mesh
Drierite™, with indicator, 10-20 mesh
Thermo Scientific Chemicals
Drierite™, with indicator, 10-20 mesh
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Packaging:
Glass Bottle
Catalog number FSA350015000
Price (MYR)
410.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
410.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDrierite™
ColorBlue
Melting Point1450.0°C
Physical FormIrregular Granules
CAS7778-18-9
View more
Adsorbent, Indicator
RUO – Research Use Only