Cumyl hydroperoxide, 80%
Cumyl hydroperoxide, 80%
Cumyl hydroperoxide, 80%
Cumyl hydroperoxide, 80%
Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%
CAS: 80-15-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA349962500
Price (MYR)
448.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
448.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCumyl hydroperoxide
Name Note80%
CAS80-15-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Heating may cause a fire.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause re
View more
Oxidation reagent
RUO – Research Use Only