Dicumyl peroxide, 99%
Dicumyl peroxide, 99%
Dicumyl peroxide, 99%
Dicumyl peroxide, 99%
Thermo Scientific Chemicals
Dicumyl peroxide, 99%
CAS: 80-43-3 | C18H22O2 | 270.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA349885000
Price (MYR)
1,076.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,076.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDicumyl peroxide
Name Note99%
CAS80-43-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Heating may cause a fire.
View more
Radical initiators
RUO – Research Use Only