3-Bromo-1-propanol, 97%
3-Bromo-1-propanol, 97%
3-Bromo-1-propanol, 97%
3-Bromo-1-propanol, 97%
Thermo Scientific Chemicals
3-Bromo-1-propanol, 97%
CAS: 627-18-9 | C3H7BrO | 138.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA349701000
Price (MYR)
2,264.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,264.00
EA
Add to cart
Specifications
Chemical Name or Material3-Bromo-1-propanol
Name Note97%
CAS627-18-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning