Sodium hydroxide, 0.2 N standard solution
Sodium hydroxide, 0.2 N standard solution
Sodium hydroxide, 0.2 N standard solution
Sodium hydroxide, 0.2 N standard solution
Thermo Scientific Chemicals
Sodium hydroxide, 0.2 N standard solution
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
Catalog number FSA349685000
Price (MYR)
238.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
238.00
EA
Add to cart
Chemical Identifiers
CAS1310-73-2, 7732-18-5
Specifications
Chemical Name or MaterialSodium hydroxide
ColorColorless
CAS1310-73-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
View more
Volumetric standard solution
RUO – Research Use Only