Celite™ 545
Thermo Scientific Chemicals
Celite™ 545
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA349675000
Price (MYR)
356.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
356.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCelite™ 545
Density336 g/L (typically)
Physical FormPowder
ColorWhite
Quantity500 g
View more
Filter aid
RUO – Research Use Only