1,2-Dibromobenzene, 98%
1,2-Dibromobenzene, 98%
1,2-Dibromobenzene, 98%
1,2-Dibromobenzene, 98%
Thermo Scientific Chemicals
1,2-Dibromobenzene, 98%
CAS: 583-53-9 | C6H4Br2 | 235.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA348931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Dibromobenzene
CAS583-53-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0