1-Aminobenzotriazole, 98%
1-Aminobenzotriazole, 98%
1-Aminobenzotriazole, 98%
1-Aminobenzotriazole, 98%
Thermo Scientific Chemicals
1-Aminobenzotriazole, 98%
CAS: 1614-12-6 | C6H6N4 | 134.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Packaging:
Glass bottle
Catalog number FSA348642500
Price (MYR)
919.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
919.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Aminobenzotriazole
Name Note98%
CAS1614-12-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning