Perchloric acid, pure, 0.1M solution in acetic acid
Perchloric acid, pure, 0.1M solution in acetic acid
Perchloric acid, pure, 0.1M solution in acetic acid
Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA348630025
Price (MYR)
1,521.00
Each
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,521.00
Each
Chemical Identifiers
CAS7601-90-3, 64-19-7
Specifications
Chemical Name or MaterialPerchloric acid
ColorColorless
Name Note0.1 M Solution in Acetic Acid
CAS7601-90-3
Health Hazard 1Danger
View more
Oxidation reagent
RUO – Research Use Only